Phosphoenolpyruvic acid monocyclohexylammonium salt ≥98.0%
Supplier: TCI America
Synonyms:
Phosphoenolpyruvic acid cyclohexylammonium salt, PEP·CHX, Phospho(enol)pyruvic acid cyclohexylammonium salt, 2-(Phosphonooxy)-2-propenoic acid monocyclohexylammonium salt
P0758-1G
P0758-100MG
TCP0758-1GEA
124.03
CAD
TCP0758-1G
TCP0758-100MG
Phosphoenolpyruvic acid monocyclohexylammonium salt ≥98.0%
Phosphoenolpyruvic acid monocyclohexylammonium salt
CAS Number: 10526-80-4
MDL Number: MFCD00036375
Molecular Formula: C3H5O6P
Molecular Weight: 267.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Storage Temperature: 0-10°C
Formula:
C₃H₅O₆P |
MDL Number:
MFCD00036375 CAS Number: 10526-80-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...