2-Hydroxyethyl phenyl sulfide ≥97.0%

Supplier: TCI America

Synonyms: 2-(Phenylthio)ethanol

P0733-25ML P0733-500ML
TCP0733-025MLEA 44.24 CAD
TCP0733-025ML TCP0733-500ML
2-Hydroxyethyl phenyl sulfide ≥97.0%
2-Hydroxyethyl phenyl sulphide

CAS Number: 699-12-7
MDL Number: MFCD00002905
Molecular Formula: C8H10OS
Molecular Weight: 154.23
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 275
Flash Point (°C): 110
Specific Gravity (20/20): 1.15


Other Product Names / Synonyms: 2-Hydroxyethyl Phenyl Sulfide

Formula: C₈H₁₀OS
MW: 154.23 g/mol
Boiling Pt: 275 °C
Density: 1.15 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00002905
CAS Number: 699-12-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b171267d30f3b3f41d756d76356a933cba7a38a&vid=4b171267d30f3b3f41d756d76356a933cba7a38a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR