Phthalazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Benzo[d]pyridazine

P0694-25G P0694-5G
TCP0694-25GEA 161.16 CAD
TCP0694-25G TCP0694-5G
Phthalazine ≥98.0% (by GC, titration analysis)
Phthalazine

CAS Number: 253-52-1
MDL Number: MFCD00006908
Molecular Formula: C8H6N2
Molecular Weight: 130.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 317
Melting point (°C): 93


Other Product Names / Synonyms: 2,3-Benzodiazine, 4,5-Benzopyridazine

Formula: C₈H₆N₂
MW: 130.15 g/mol
Boiling Pt: 317 °C
Melting Pt: 93 °C
MDL Number: MFCD00006908
CAS Number: 253-52-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65539f9e111a1ee2cdd82f754fb28a87a14f5003&vid=65539f9e111a1ee2cdd82f754fb28a87a14f5003&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR