Polyethylene glycol monododecyl ether, M.W. n=∼25

Supplier: TCI America

Synonyms: Polidocanol

P0671-500G P0671-25G
TCP0671-500GEA 61.15 CAD
TCP0671-500G TCP0671-025G
Polyethylene glycol monododecyl ether, M.W. n=∼25
Polyethylene glycol monododecyl ether

CAS Number: 9002-92-0
MDL Number: MFCD00080891
Form: Crystal
Color: White
Flash Point (°C): 170


Other Product Names / Synonyms: Polyethylene Glycol Monolauryl Ether

Formula: HO(CH2CH2O)nC12H25
Flash Pt: 170 °C
MDL Number: MFCD00080891
CAS Number: 9002-92-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=803363c0a157e63aa93800183cbf4f80812026ec&vid=51f22d3f59fd146f0e3fcb02da81271e06aeb1f5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR