Poly(vinyl formal)
Supplier: TCI America
Synonyms:
Polyvinylformal, Formvar
P0641-500G
P0641-25G
TCP0641-500GEA
259.12
CAD
TCP0641-500G
TCP0641-025G
Poly(vinyl formal)
Poly(vinyl formal)
CAS Number: 63450-15-7
MDL Number: MFCD00084463
Form: Grain
Color: Slightly Pale Yellow
Product Categories: Materials Chemistry, Polymers
Formula:
(C₃H₆O₂)n |
MDL Number:
MFCD00084463 CAS Number: 63450-15-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...