2-(Pyridin-3-yl)acetic acid hydrochloride ≥99.0%

Supplier: TCI America

Synonyms: Pyridyl-3-acetic acid hydrochloride, 3-Pyridylacetic acid hydrochloride

P0639-25G P0639-10G
TCP0639-25GEA 266.07 CAD
TCP0639-25G TCP0639-010G
2-(Pyridin-3-yl)acetic acid hydrochloride ≥99.0%
2-(Pyridin-3-yl)acetic acid hydrochloride

CAS Number: 6419-36-9
MDL Number: MFCD00012819
Molecular Formula: C7H7NO2
Molecular Weight: 173.60
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 160


Other Product Names / Synonyms: 3-Pyridineacetic Acid Hydrochloride

Formula: C₇H₈ClNO₂
MW: 173.60 g/mol
Melting Pt: 160 °C
MDL Number: MFCD00012819
CAS Number: 6419-36-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9e523d81a38e1e5150a8c50cb43f1dd8ada682a&vid=a9e523d81a38e1e5150a8c50cb43f1dd8ada682a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR