Polyvinylpyrrolidone, M.W. K 90 (MW ∼ 360,000)

Supplier: TCI America

P0473-25G P0473-500G
TCP0473-025GEA 64.54 CAD
TCP0473-025G TCP0473-500G
Polyvinylpyrrolidone, M.W. K 90 (MW ∼ 360,000)
Polyvinylpyrrolidone

CAS Number: 9003-39-8
Form: Crystals
Color: White


Product Categories: Materials Chemistry, Polymer Research Reagents

Formula: (C₆H₉NO)n
MDL Number: MFCD01076626
CAS Number: 9003-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR