Polyvinyl alcohol, M.W. n=1750±50

Supplier: TCI America

P0469-25G P0469-500G
TCP0469-25GEA 74.42 CAD
TCP0469-25G TCP0469-500G
Polyvinyl alcohol, M.W. n=1750±50
Polyvinyl alcohol

CAS Number: 9002-89-5
Form: Crystals
Color: White
Flash Point (°C): 77


Product Categories: Materials Chemistry, Polymer Research Reagents

Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Solubility in hot Water almost transparency
Drying loss max. 7.0 %
degree's of saponification 97.0 to 100.0 mol%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR