Piperine ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

P0460-5G P0460-1G
TCP0460-005GEA 90.03 CAD
TCP0460-005G TCP0460-001G
Piperine ≥97.0% (by HPLC, total nitrogen)
Piperine

CAS Number: 94-62-2
MDL Number: MFCD00005839
Molecular Formula: C17H19NO3
Molecular Weight: 285.34
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Crystal
Melting point (°C): 129


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Alkaloids (Phytochemicals), Piperidine Alkaloids
Other Product Names / Synonyms: 1-[5-(1,3-Benzodioxol-5-yl)-2,4-pentadienoyl]piperidine, 1-Piperoylpiperidine, 1-Piperylpiperidine

Formula: C₁₇H₁₉NO₃
Melting Pt: 129 °C
MDL Number: MFCD00005839
CAS Number: 94-62-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f5c16fb2aaeeefeabaac6bc8d714c034008b5b8&vid=3f5c16fb2aaeeefeabaac6bc8d714c034008b5b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR