Polyethyleneimine ∼30% in water
Supplier: TCI America
P0381-500G
P0381-25G
TCP0381-500GEA
110.76
CAD
TCP0381-500G
TCP0381-25G
Polyethyleneimine ∼30% in water
Polyethyleneimine
CAS Number: 9002-98-6
MDL Number: MFCD00084427
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.05
Other Product Names / Synonyms: Ethyleneimine Polymer
Formula:
(CH2CH2NH)n Density: 1.05 (20 °C) |
MDL Number:
MFCD00084427 CAS Number: 9002-98-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...