Sodium-1-propanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P0347-25G P0347-5G
TCP0347-025GEA 317.42 CAD
TCP0347-025G TCP0347-005G
Sodium-1-propanesulfonate ≥98.0% (by titrimetric analysis)
Sodium-1-propanesulfonate

CAS Number: 14533-63-2
MDL Number: MFCD00062546
Molecular Formula: C3H8O3S
Molecular Weight: 146.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 1-Propanesulfonic Acid Sodium Salt

Formula: C₃H₈O₃S
MDL Number: MFCD00062546
CAS Number: 14533-63-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26f34d67207fab8033c3e0d113ec54b0f3a2dbef&vid=26f34d67207fab8033c3e0d113ec54b0f3a2dbef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR