L-Proline methyl ester hydrochloride ≥98.0% (by total nitrogen basis)

Supplier: TCI America

P0342-5G P0342-25G
TCP0342-005GEA 97.96 CAD
TCP0342-005G TCP0342-025G
L-Proline methyl ester hydrochloride ≥98.0% (by total nitrogen basis)
L-Proline methyl ester hydrochloride

CAS Number: 2133-40-6
MDL Number: MFCD00012708
Molecular Formula: C6H11NO2
Molecular Weight: 165.62
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Boiling point (°C): 55
Melting point (°C): 73
Specific rotation [a]20/D: -31.5 deg (C=1, H2O)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Pro-OMe.HCl

Formula: C₆H₁₁NO₂·HCl
Boiling Pt: 55 °C
Melting Pt: 73 °C
MDL Number: MFCD00012708
CAS Number: 2133-40-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3dea729d85c69b8a209c5195526d786a546a2571&vid=3dea729d85c69b8a209c5195526d786a546a2571&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR