1-Phenyl-2-pyrrolidone ≥99.0%

Supplier: TCI America

Synonyms: 1-Phenyl-2-pyrrolidinone

P0319-25G P0319-5G
TCP0319-025GEA 139.04 CAD
TCP0319-025G 76220-192
1-Phenyl-2-pyrrolidone ≥99.0%
1-Phenyl-2-pyrrolidone

CAS Number: 4641-57-0
MDL Number: MFCD00003192
Molecular Formula: C10H11NO
Molecular Weight: 161.20
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 68

Formula: C₁₀H₁₁NO
MW: 161.20 g/mol
Melting Pt: 68 °C
MDL Number: MFCD00003192
CAS Number: 4641-57-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a912fca9e3af43cbd72dd9da8ef457a628650a95&vid=a912fca9e3af43cbd72dd9da8ef457a628650a95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR