Di-n-octyl phthalate ≥98.0%
Supplier: TCI America
P0304-25ML
P0304-500ML
TCP0304-025MLEA
67.5
CAD
TCP0304-025ML
TCP0304-500ML
Di-n-octyl phthalate ≥98.0%
Di-n-octyl phthalate
CAS Number: 117-84-0
MDL Number: MFCD00015292
Molecular Formula: C24H38O4
Molecular Weight: 390.56
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 380
Flash Point (°C): 219
Specific Gravity (20/20): 0.98
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Phthalic Acid Di-n-octyl Ester
Formula:
C₂₄H₃₈O₄ Boiling Pt: 380 °C Density: 0.98 (20 °C) Flash Pt: 219 °C |
MDL Number:
MFCD00015292 CAS Number: 117-84-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...