Phenyl phosphate disodium salt ≥98.0%

Supplier: TCI America

Synonyms: Phosphoric acid phenyl ester disodium salt, Disodium phenyl phosphate, Phosphoric acid monophenyl ester disodium salt

P0208-25G P0208-500G
TCP0208-025GEA 105.86 CAD
TCP0208-025G TCP0208-500G
Phenyl phosphate disodium salt ≥98.0%
Phenyl phosphate disodium salt

CAS Number: 3279-54-7 MDL Number: MFCD00002133 Molecular Formula: C6H7O4P Molecular Weight: 218.06 Purity/Analysis Method: 98.0% (T) Form: Crystal


Form: Crystal


Other Product Names / Synonyms: Phosphoric Acid Phenyl Ester Disodium Salt

Formula: C₆H₅Na₂O₄P·xH₂O
MW: 218.06 g/mol (anhy)
MDL Number: MFCD00002133
CAS Number: 3279-54-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=685bd3ed2e4013da2e79f787ca9a2d5f83ebb8f8&vid=685bd3ed2e4013da2e79f787ca9a2d5f83ebb8f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR