N-Phenylglycine ≥97.0% (by titrimetric analysis)

Supplier: TCI America

P0180-25G P0180-100G
TCP0180-025GEA 93.73 CAD
TCP0180-025G TCP0180-100G
N-Phenylglycine ≥97.0% (by titrimetric analysis)
N-Phenylglycine

CAS Number: 103-01-5
MDL Number: MFCD00014009
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 120


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: Anilinoacetic Acid

Formula: C₈H₉NO₂
Melting Pt: 120 °C
MDL Number: MFCD00014009
CAS Number: 103-01-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526621d9c3df57dc0b2a597af4f06e3e214d7dac&vid=526621d9c3df57dc0b2a597af4f06e3e214d7dac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR