N-Benzoyl-N-phenylhydroxylamine ≥98.0%
Supplier: TCI America
Synonyms:
Tantalon, N-Phenylbenzohydroxamic Acid, N-Hydroxy-N-phenylbenzamide
P0158-5G
P0158-25G
TCP0158-005GEA
127.02
CAD
TCP0158-005G
TCP0158-025G
N-Benzoyl-N-phenylhydroxylamine ≥98.0%
N-Benzoyl-N-phenylhydroxylamine
CAS Number: 304-88-1
MDL Number: MFCD00002111
Molecular Formula: C13H11NO2
Molecular Weight: 213.24
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 122
Product Categories: Analytical Science, Bipyridyls, etc. (Chelating Agents)
Other Product Names / Synonyms: N-Phenylbenzohydroxamic Acid, Tantalon
Formula:
C₁₃H₁₁NO₂ MW: 213.24 g/mol Melting Pt: 122 °C |
MDL Number:
MFCD00002111 CAS Number: 304-88-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...