4-(Phenylazo)diphenylamine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
4-Phenylazodiphenylamine
P0146-5G
P0146-25G
TCP0146-005GEA
63.2
CAD
TCP0146-005G
TCP0146-025G
4-(Phenylazo)diphenylamine ≥98.0% (by titrimetric analysis)
4-(Phenylazo)diphenylamine
CAS Number: 101-75-7
MDL Number: MFCD00003023
Molecular Formula: C18H15N3
Molecular Weight: 273.34
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 86
Other Product Names / Synonyms: 4-Anilinoazobenzene
Formula:
C₁₈H₁₅N₃ MW: 273.34 g/mol Melting Pt: 86 °C |
MDL Number:
MFCD00003023 CAS Number: 101-75-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...