Sodium-4-hydroxybenzenesulfonate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

P0105-25G P0105-500G
TCP0105-025GEA 36.34 CAD
TCP0105-025G TCP0105-500G
Sodium-4-hydroxybenzenesulfonate ≥98.0% (by HPLC, titration analysis)
Sodium-4-hydroxybenzenesulfonate

CAS Number: 825-90-1
MDL Number: MFCD00044734
Molecular Formula: C6H6O4S
Molecular Weight: 196.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 4-Hydroxybenzenesulfonic Acid Sodium Salt, p-Phenolsulfonic Acid Sodium Salt, Sodium p-Phenolsulfonate

Formula: C₆H₆O₄S
MDL Number: MFCD00044734
CAS Number: 825-90-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e4ab15d31b1f10dacd2502c03fcd4ba9641a564&vid=0e4ab15d31b1f10dacd2502c03fcd4ba9641a564&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR