Penta-O-acetyl-β-D-glucopyranose ≥99.0%

Supplier: TCI America

Synonyms: β-D-Glucose Pentaacetate, Pentaacetyl-β-D-glucose, B-D-Glucose Pentaacetate, (2S,3R,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate

P0028-500G P0028-100G
TCP0028-500GEA 274.45 CAD
TCP0028-500G TCP0028-100G
Penta-O-acetyl-β-D-glucopyranose ≥99.0%
Penta-O-acetyl-beta-D-glucopyranose

CAS Number: 604-69-3
MDL Number: MFCD00006597
Molecular Formula: C16H22O11
Molecular Weight: 390.34
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 132
Specific rotation [a]20/D: 4.5 deg (C=5, CHCl3)


Product Categories: Bioscience, Glucose, O-Substituted Sugars, Cosmetic Ingredients for Research and Experimental Use
Other Product Names / Synonyms: beta-D-Glucose Pentaacetate, Pentaacetyl-beta-D-glucose

Formula: C₁₆H₂₂O₁₁
MW: 390.34 g/mol
Melting Pt: 132 °C
MDL Number: MFCD00006597
CAS Number: 604-69-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ea9995424aacd868cad4180cc11cce06bccc8b5&vid=9ea9995424aacd868cad4180cc11cce06bccc8b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR