N-Octyl-β-D-glucopyranoside ≥96.0% for biochemical research

Supplier: TCI America

Synonyms: Octylglucopyranoside1-O-n-Octyl-β-D-glucopyranosideOctyl-β-D-glucopyranosiden-Octyl β-D-glucopyranosideOG(2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol

O0355-1G
TCO0355-1GEA 180.12 CAD
TCO0355-1G
N-Octyl-β-D-glucopyranoside ≥96.0% for biochemical research
N-Octyl-β-D-glucopyranoside

[for Biochemical Research]
CAS Number: 29836-26-8
MDL Number: MFCD00063288
Molecular Formula: C14H28O6
Molecular Weight: 292.37
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Specific rotation [a]20/D: -27 deg (C=1, MeOH)

Formula: C₁₄H₂₈O₆
MW: 292.37 g/mol
MDL Number: MFCD00063288
CAS Number: 29836-26-8
https://ca.vwr.com/stibo/web/std.lang.all/05/87/28990587.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR