2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol ≥98.0%

Supplier: TCI America

O0294-25G O0294-5G
TCO0294-25GEA 289.39 CAD
TCO0294-25G TCO0294-5G
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol ≥98.0%

CAS Number: 355-74-8
MDL Number: MFCD00069087
Molecular Formula: C6H6F8O2
Molecular Weight: 262.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 68


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,6-Dihydroxy-2,2,3,3,4,4,5,5-octafluorohexane

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR