Chromone-3-carboxaldehyde ≥98.0%

Supplier: TCI America

Synonyms: 4-Oxo-4H-chromene-3-carbaldehyde

O0291-25G O0291-5G
TCO0291-25GEA 128.93 CAD
TCO0291-25G TCO0291-5G
Chromone-3-carboxaldehyde ≥98.0%
Chromone-3-carboxaldehyde

CAS Number: 17422-74-1
MDL Number: MFCD00014667
Molecular Formula: C10H6O3
Molecular Weight: 174.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 153


Other Product Names / Synonyms: 3-Formylchromone, 4-Oxo-4H-1-benzopyran-3-carboxaldehyde

Formula: C₁₀H₆O₃
MW: 174.16 g/mol
Melting Pt: 153 °C
MDL Number: MFCD00014667
CAS Number: 17422-74-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2eda207c51e034f27a94e234b91b0e6159e1610a&vid=2eda207c51e034f27a94e234b91b0e6159e1610a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR