Chromone-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

O0290-25G O0290-5G
TCO0290-25GEA 999.98 CAD
TCO0290-25G TCO0290-5G
Chromone-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Chromone-2-carboxylic Acid

CAS Number: 4940-39-0
MDL Number: MFCD00006838
Molecular Formula: C10H6O4
Molecular Weight: 190.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 4-Oxo-4H-1-benzopyran-2-carboxylic Acid

Formula: C₁₀H₆O₄
MDL Number: MFCD00006838
CAS Number: 4940-39-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cad1685afcf07e276a399eab9e30652e1daec746&vid=cad1685afcf07e276a399eab9e30652e1daec746&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR