4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl ≥95.0% (by GC, titration analysis) free radical

Supplier: TCI America

Synonyms: TEMPONE, 4-Oxo-2,2,6,6-tetramethylpiperidinooxy, 4-Oxo-TEMPO

O0266-25G O0266-5G
TCO0266-25GEA 524.56 CAD
TCO0266-25G TCO0266-005G
4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl ≥95.0% (by GC, titration analysis) free radical
4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl

CAS Number: 2896-70-0
MDL Number: MFCD00006187
Molecular Formula: C9H16NO2
Molecular Weight: 170.23
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 37
Storage Temperature: <0°C


Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Precise Polymerization Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Oxo-TEMPO

Formula: C₉H₁₆NO₂
MW: 170.23 g/mol
Melting Pt: 37 °C
MDL Number: MFCD00006187
CAS Number: 2896-70-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aef28fdb4c38376390cf7542e814e1b351299b97&vid=aef28fdb4c38376390cf7542e814e1b351299b97&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR