Methyl trans-9-octadecenoate ≥98.0%
Supplier: TCI America
Synonyms:
Elaidic Acid Methyl Ester, trans-9-Octadecenoic Acid Methyl Ester, (E)-Methyl octadec-9-enoate, Methyl Elaidate
CAS Number: 1937-62-8
MDL Number: MFCD00066521
Molecular Formula: C19H36O2
Molecular Weight: 296.50
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Specific Gravity (20/20): 0.87
Storage Temperature: 0-10°C
Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Elaidic Acid Methyl Ester, Methyl Elaidate, trans-9-Octadecenoic Acid Methyl Ester
Formula:
C₁₉H₃₆O₂ MW: 296.50 g/mol Boiling Pt: 220 °C Density: 0.87 (20 °C) |
MDL Number:
MFCD00066521 CAS Number: 1937-62-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...