Dimethyl (2-oxopropyl)phosphonate ≥95.0%
Supplier: TCI America
Synonyms:
Dimethyl acetylmethylphosphonate
CAS Number: 4202-14-6
MDL Number: MFCD00008769
Molecular Formula: C5H11O4P
Molecular Weight: 166.11
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 124
Flash Point (°C): 93
Specific Gravity (20/20): 1.21
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Acetonylphosphonic Acid Dimethyl Ester, Dimethyl Acetonylphosphonate, (2-Oxopropyl)phosphonic Acid Dimethyl Ester
Formula:
C₅H₁₁O₄P MW: 166.12 g/mol Boiling Pt: 124 °C Density: 1.21 (20 °C) Flash Pt: 93 °C |
MDL Number:
MFCD00008769 CAS Number: 4202-14-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...