Sodium-4-n-octylbenzenesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

O0203-5G O0203-25G
TCO0203-5GEA 339.7 CAD
TCO0203-5G TCO0203-25G
Sodium-4-n-octylbenzenesulfonate ≥98.0% (by titrimetric analysis)
Sodium-4-n-octylbenzenesulfonate

CAS Number: 6149-03-7
MDL Number: MFCD00007512
Molecular Formula: C14H22O3S
Molecular Weight: 292.37
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 300


Other Product Names / Synonyms: 4-n-Octylbenzenesulfonic Acid Sodium Salt

Formula: C₁₄H₂₂O₃S
Melting Pt: 300 °C
MDL Number: MFCD00007512
CAS Number: 6149-03-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2383976ef2e1a3ccab0e65e2c13be3f6e29ae48e&vid=2383976ef2e1a3ccab0e65e2c13be3f6e29ae48e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR