Potassium oxonate ≥98.0%

Supplier: TCI America

Synonyms: Potassium Allantoxanate, Oxonic acid Potassium Salt, Potassium 4,6-Dihydroxy-1,3,5-triazine-2-carboxylate, Allantoxanic acid Potassium Salt, Potassium 4,6-dioxo-1,4,5,6-tetrahydro-1,3,5-triazine-2-carboxylate

O0164-5G O0164-25G
TCO0164-005GEA 48.25 CAD
TCO0164-005G TCO0164-025G
Potassium oxonate ≥98.0%
Potassium oxonate

CAS Number: 2207-75-2
MDL Number: MFCD00010565
Molecular Formula: C4H3N3O4
Molecular Weight: 195.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 300


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Reagents for Life-style related Disease Models
Other Product Names / Synonyms: Allantoxanic Acid Potassium Salt, Oxonic Acid Potassium Salt, Potassium Allantoxanate, Potassium 4,6-Dihydroxy-1,3,5-triazine-2-carboxylate

Formula: C₄H₃N₃O₄
MW: 195.18 g/mol
Melting Pt: 300 °C
MDL Number: MFCD00010565
CAS Number: 2207-75-2

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR