Diethyl oxalacetate sodium salt ≥95.0% (by titrimetric analysis)

Supplier: TCI America

O0074-25G O0074-500G
TCO0074-025GEA 113.76 CAD
TCO0074-025G TCO0074-500G
Diethyl oxalacetate sodium salt ≥95.0% (by titrimetric analysis)
Diethyl oxalacetate sodium salt

CAS Number: 40876-98-0
MDL Number: MFCD00035571
Molecular Formula: C8H12O5
Molecular Weight: 210.16
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Other Product Names / Synonyms: Oxalacetic Acid Diethyl Ester Sodium Salt, Sodium Diethyl Oxalacetate

Formula: C₈H₁₂O₅
MDL Number: MFCD00035571
CAS Number: 40876-98-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc1811f58c54b0992dff8e9e2ec6da59c9d6c01e&vid=dc1811f58c54b0992dff8e9e2ec6da59c9d6c01e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR