trans,cis-2,6-Nonadienal ≥95.0%

Supplier: TCI America

N0836-5G N0836-1G
TCN0836-5GEA 336.54 CAD
TCN0836-5G TCN0836-1G
trans,cis-2,6-Nonadienal ≥95.0%
trans,cis-2,6-Nonadienal

CAS Number: 557-48-2
MDL Number: MFCD00007009
Molecular Formula: C9H14O
Molecular Weight: 138.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 83
Specific Gravity (20/20): 0.87

Formula: C₉H₁₄O
MW: 138.21 g/mol
Boiling Pt: 90 °C
Density: 0.87 (20 °C)
Flash Pt: 83 °C
MDL Number: MFCD00007009
CAS Number: 557-48-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=badcb36f4a689074f284a59f8ca308f6e39d7bbe&vid=badcb36f4a689074f284a59f8ca308f6e39d7bbe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR