3-Nitrosalicylaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America

N0823-1G N0823-5G
TCN0823-1GEA 117.39 CAD
TCN0823-1G TCN0823-5G
3-Nitrosalicylaldehyde ≥98.0% (by GC, titration analysis)
3-Nitrosalicylaldehyde

CAS Number: 5274-70-4
MDL Number: MFCD00041874
Molecular Formula: C7H5NO4
Molecular Weight: 167.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 110


Other Product Names / Synonyms: 2-Hydroxy-3-nitrobenzaldehyde

Formula: C₇H₅NO₄
Melting Pt: 110 °C
MDL Number: MFCD00041874
CAS Number: 5274-70-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18a2813d881285b07bb777cf52e549dfe910af12&vid=c9ed58cf571ac0ed35abd9cd7c51d59c95076049&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR