3'-Nitropropiophenone ≥98.0%

Supplier: TCI America

Synonyms: m-Nitropropiophenone, 1-(3-Nitrophenyl)propan-1-one

N0789-5G N0789-25G
TCN0789-5GEA 257.54 CAD
TCN0789-5G TCN0789-25G
3'-Nitropropiophenone ≥98.0%
3'-Nitropropiophenone

CAS Number: 17408-16-1
MDL Number: MFCD00051516
Molecular Formula: C9H9NO3
Molecular Weight: 179.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 100


Other Product Names / Synonyms: 3-Nitrophenyl Ethyl Ketone

Formula: C₉H₉NO₃
Melting Pt: 100 °C
MDL Number: MFCD00051516
CAS Number: 17408-16-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=711ddc37a0374ce4fe95d099e04ac029df7d7ff4&vid=711ddc37a0374ce4fe95d099e04ac029df7d7ff4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR