1-(2-Naphthyl)ethanol ≥98.0%

Supplier: TCI America

N0788-25G N0788-5G
TCN0788-25GEA 290.88 CAD
TCN0788-25G TCN0788-5G
1-(2-Naphthyl)ethanol ≥98.0%
1-(2-Naphthyl)ethanol

CAS Number: 7228-47-9
MDL Number: MFCD00004111
Molecular Formula: C12H12O
Molecular Weight: 172.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 170
Melting point (°C): 76


Other Product Names / Synonyms: alpha-Methyl-2-naphthalenemethanol, 2-(1-Hydroxyethyl)naphthalene

Formula: C₁₂H₁₂O
Boiling Pt: 170 °C
Melting Pt: 76 °C
MDL Number: MFCD00004111
CAS Number: 7228-47-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6ae5b17c26c42d786427f16f5e83f57a64718c9&vid=d6ae5b17c26c42d786427f16f5e83f57a64718c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR