4-Nitrothioanisole ≥98.0%

Supplier: TCI America

N0761-25G N0761-5G
TCN0761-25GEA 368.14 CAD
TCN0761-25G TCN0761-5G
4-Nitrothioanisole ≥98.0%
4-Nitrothioanisole

CAS Number: 701-57-5
Molecular Formula: C7H7NO2S
Molecular Weight: 169.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 71


Other Product Names / Synonyms: 1-(Methylthio)-4-nitrobenzene

Formula: C₇H₇NO₂S
Boiling Pt: 140 °C
Melting Pt: 71 °C
MDL Number: MFCD00010868
CAS Number: 701-57-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2dfc8b35ed4a06dd0f8b15670ca079aa1e0e39&vid=ec2dfc8b35ed4a06dd0f8b15670ca079aa1e0e39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR