Naphthalene-1,4,5,8-tetracarboxylic dianhydride ≥98.0% (by HPLC), purified by sublimation

Supplier: TCI America

N0755-1G N0755-5G
TCN0755-1GEA 195.92 CAD
TCN0755-1G TCN0755-5G
Naphthalene-1,4,5,8-tetracarboxylic dianhydride ≥98.0% (by HPLC), purified by sublimation
Naphthalene-1,4,5,8-tetracarboxylic dianhydride

CAS Number: 81-30-1
MDL Number: MFCD00006915
Molecular Formula: C14H4O6
Molecular Weight: 268.18
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Product Categories: Materials Chemistry, Organic Transistor Materials, Conducting Polymer Research Reagents, Monomers, Polyimide Raw Materials, Highly Purified Reagents
Other Product Names / Synonyms: NTCDA

Formula: C₁₄H₄O₆
MDL Number: MFCD00006915
CAS Number: 81-30-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0788a1c8679e2f763050d33a467bf02755ef3de&vid=a4c304e5acb2cfe54b7ead00d982c96e003be6b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR