(S)-(-)-1-(2-Naphthyl)ethylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (S)-1-(Naphthalen-2-yl)ethanamine

N0726-5G N0726-1G
TCN0726-5GEA 330.22 CAD
TCN0726-5G TCN0726-1G
(S)-(-)-1-(2-Naphthyl)ethylamine ≥98.0% (by GC, titration analysis)
(S)-(-)-1-(2-Naphthyl)ethylamine

CAS Number: 3082-62-0
MDL Number: MFCD00085366
Molecular Formula: C12H13N
Molecular Weight: 171.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 143
Melting point (°C): 53
Flash Point (°C): 100
Specific rotation [a]20/D: -20 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-2-(1-Aminoethyl)naphthalene

Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Boiling Pt: 143 °C
Melting Pt: 53 °C
Flash Pt: 100 °C
MDL Number: MFCD00085366
CAS Number: 3082-62-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72ee959d6485cc3172801e4dd86a17dec780b6d2&vid=72ee959d6485cc3172801e4dd86a17dec780b6d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR