(R)-(+)-1-(2-Naphthyl)ethylamine ≥98.0%

Supplier: TCI America

Synonyms: (R)-1-(2-Naphthyl)ethylamine

N0724-5G N0724-1G
TCN0724-5GEA 448.72 CAD
TCN0724-5G TCN0724-1G
(R)-(+)-1-(2-Naphthyl)ethylamine ≥98.0%
(R)-(+)-1-(2-Naphthyl)ethylamine

CAS Number: 3906-16-9
MDL Number: MFCD00085367
Molecular Formula: C12H13N
Molecular Weight: 171.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 166
Melting point (°C): 53
Flash Point (°C): 100
Specific rotation [a]20/D: 22 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-2-(1-Aminoethyl)naphthalene

Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Boiling Pt: 166 °C
Melting Pt: 53 °C
Flash Pt: 100 °C
MDL Number: MFCD00085367
CAS Number: 3906-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=456e9befccf324c163261a1f21787466bd2e3f00&vid=456e9befccf324c163261a1f21787466bd2e3f00&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR