2-Nitro-α,α,α-trifluoro-p-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

N0720-5G N0720-25G
TCN0720-5GEA 64.78 CAD
TCN0720-5G TCN0720-25G
2-Nitro-α,α,α-trifluoro-p-toluic acid ≥98.0% (by GC, titration analysis)
2-Nitro-α,α,α-trifluoro-p-toluic acid

CAS Number: 320-94-5
Molecular Formula: C8H4F3NO4
Molecular Weight: 235.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 138

Formula: C₈H₄F₃NO₄
Melting Pt: 138 °C
MDL Number: MFCD00007142
CAS Number: 320-94-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d114e168b3c178d041cfeea60e4776fab3f707f3&vid=d114e168b3c178d041cfeea60e4776fab3f707f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR