(1-Naphthylmethyl)triphenylphosphonium chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
N0700-5G
N0700-25G
TCN0700-5GEA
83.74
CAD
TCN0700-5G
TCN0700-25G
(1-Naphthylmethyl)triphenylphosphonium chloride ≥98.0% (by titrimetric analysis)
(1-Naphthylmethyl)triphenylphosphonium chloride
CAS Number: 23277-00-1
MDL Number: MFCD00031658
Molecular Formula: C29H24ClP
Molecular Weight: 438.93
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Triphenyl(1-naphthylmethyl)phosphonium Chloride
Formula:
C₂₉H₂₄ClP |
MDL Number:
MFCD00031658 CAS Number: 23277-00-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...