4-Nitrobenzenesulfonamide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

N0697-25G N0697-5G
TCN0697-25GEA 238.58 CAD
TCN0697-25G TCN0697-5G
4-Nitrobenzenesulfonamide ≥98.0% (by HPLC, titration analysis)
4-Nitrobenzenesulphonamide

CAS Number: 6325-93-5
MDL Number: MFCD00007937
Molecular Formula: C6H6N2O4S
Molecular Weight: 202.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 182

Formula: C₆H₆N₂O₄S
MW: 202.19 g/mol
Melting Pt: 182 °C
MDL Number: MFCD00007937
CAS Number: 6325-93-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95c6ce21ff2da9b286d932a03a7c2fa830f49914&vid=95c6ce21ff2da9b286d932a03a7c2fa830f49914&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR