Perfluoro-tert-butanol ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Perfluoro-t-butanol, Nonafluoro-tert-butyl alcohol, 1,1,1,3,3,3-Hexafluoro-2-trifluoromethyl-2-propanol, Perfluoro-tert-butyl alcohol

N0692-1G N0692-5G N0692-25G
TCN0692-1GEA 99.54 CAD
TCN0692-1G TCN0692-5G TCN0692-25G
Perfluoro-tert-butanol ≥98.0% (by GC)
Perfluoro-tert-butanol

CAS Number: 2378-02-1 MDL Number: MFCD00042092 Molecular Formula: C4HF9O Molecular Weight: 236.04 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 45 Specific Gravity (20/20): 1.70


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks Other Product Names / Synonyms: 1,1,1,3,3,3-Hexafluoro-2-trifluoromethyl-2-propanol, Perfluoro-tert-butanol, Nonafluoro-tert-butanol, Perfluoro-tert-butyl Alcohol

Formula: C₄HF₉O
MW: 236.04 g/mol
Boiling Pt: 45 °C
Density: 1.70 (20 °C)
MDL Number: MFCD00042092
CAS Number: 2378-02-1
UN: 2810
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8c1b1ffcd781d60932f1ce206db7777b3eab1fd&vid=c8c1b1ffcd781d60932f1ce206db7777b3eab1fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Specific gravity (20/20) 1.6950 to 1.7050

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR