4-Nitrocinnamyl alcohol ≥98.0%

Supplier: TCI America

N0690-1G N0690-5G
TCN0690-1GEA 121.66 CAD
TCN0690-1G TCN0690-5G
4-Nitrocinnamyl alcohol ≥98.0%
4-Nitrocinnamyl alcohol

CAS Number: 1504-63-8
MDL Number: MFCD00017045
Molecular Formula: C9H9NO3
Molecular Weight: 179.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 128

Formula: C₉H₉NO₃
Melting Pt: 128 °C
MDL Number: MFCD00017045
CAS Number: 1504-63-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53d75cf0fa9ad9e42a0c99c51c4e0001448955ec&vid=53d75cf0fa9ad9e42a0c99c51c4e0001448955ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR