7-Nitroindole ≥98.0%

Supplier: TCI America

N0685-1G N0685-5G
TCN0685-1GEA 86.9 CAD
TCN0685-1G TCN0685-5G
7-Nitroindole ≥98.0%
7-Nitroindole

CAS Number: 6960-42-5
MDL Number: MFCD00005683
Molecular Formula: C8H6N2O2
Molecular Weight: 162.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 97

Formula: C₈H₆N₂O₂
Melting Pt: 97 °C
MDL Number: MFCD00005683
CAS Number: 6960-42-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47076bbe44188350bee2432e6b26d89ad131135b&vid=47076bbe44188350bee2432e6b26d89ad131135b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR