4-Nitro-L-phenylalanine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: H-Phe(4-NO2)-OH, (S)-4-Nitrophenylalanine, (S)-2-Amino-3-(4-nitrophenyl)propanoic acid

N0682-1G N0682-5G
TCN0682-1GEA 33.18 CAD
TCN0682-1G TCN0682-5G
4-Nitro-L-phenylalanine ≥98.0% (by HPLC, titration analysis)
4-Nitro-L-phenylalanine

CAS Number: 949-99-5
MDL Number: MFCD00150543
Molecular Formula: C9H10N2O4
Molecular Weight: 210.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: 6.0 deg (C=1, 1mol/L HCl)


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Phe(4-NO2)-OH

Formula: C₉H₁₀N₂O₄
MW: 210.19 g/mol
MDL Number: MFCD00051221
CAS Number: 949-99-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93a1b276184be3ad0eae0604662cc4be843e569d&vid=93a1b276184be3ad0eae0604662cc4be843e569d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR