D-Norvaline ≥99.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
H-D-Nva-OH, (R)-(-)-Norvaline, D(-)-Norvaline
N0673-5G
N0673-1G
N0673-25G
TCN0673-5GEA
524.56
CAD
TCN0673-5G
TCN0673-1G
TCN0673-25G
D-Norvaline ≥99.0% (by titrimetric analysis)
D-Norvaline
CAS Number: 2013-12-9
MDL Number: MFCD00008097
Molecular Formula: C5H11NO2
Molecular Weight: 117.15
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Specific rotation [a]20/D: -25 deg (C=10, 6mol/L HCl)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (R)-2-Aminovaleric Acid
Formula:
C₅H₁₁NO₂ MW: 117.15 g/mol |
MDL Number:
MFCD00008097 CAS Number: 2013-12-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...