2-(1-Naphthalene)ethanol ≥95.0%

Supplier: TCI America

Synonyms: 2-(Naphthalen-1-yl)ethanol, 2-(1-Naphthyl)ethanol

N0575-25G N0575-5G
TCN0575-025GEA 401.32 CAD
TCN0575-025G TCN0575-005G
2-(1-Naphthalene)ethanol ≥95.0%
2-(1-Naphthalene)ethanol

CAS Number: 773-99-9
MDL Number: MFCD00004050
Molecular Formula: C12H12O
Molecular Weight: 172.23
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 179
Melting point (°C): 60


Other Product Names / Synonyms: 1-(2-Hydroxyethyl)naphthalene, 2-(1-Naphthalene)ethanol

Formula: C₁₂H₁₂O
Boiling Pt: 179 °C
Melting Pt: 60 °C
MDL Number: MFCD00004050
CAS Number: 773-99-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2e7e33075b4562f77f331e71a883a13a12b5b18&vid=a2e7e33075b4562f77f331e71a883a13a12b5b18&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR