4'-Nitrobenzo-18-crown-6 (4'-nitrobenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥97.0%

Supplier: TCI America

N0562-1G N0562-5G
TCN0562-001GEA 246.48 CAD
TCN0562-001G TCN0562-005G
4'-Nitrobenzo-18-crown-6 (4'-nitrobenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥97.0%
4'-Nitrobenzo-18-crown-6 (4'-Nitrobenzo-1,4,7,10,13,16-hexoxacyclooctadecane)

CAS Number: 53408-96-1
MDL Number: MFCD00143207
Molecular Formula: C16H23NO8
Molecular Weight: 357.36
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 86


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 2,3-(4-Nitrobenzo)-1,4,7,10,13,16-hexaoxacyclooctadec-2-ene

Formula: C₁₆H₂₃NO₈
Melting Pt: 86 °C
MDL Number: MFCD00143207
CAS Number: 53408-96-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f71995343f4e8da3bc8c96a0b0d63525b49ca9d&vid=9f71995343f4e8da3bc8c96a0b0d63525b49ca9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR