4'-Nitrobenzo-15-crown-5 (4'-nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥99.0%

Supplier: TCI America

N0561-1G N0561-5G
TCN0561-001GEA 114.08 CAD
TCN0561-001G TCN0561-005G
4'-Nitrobenzo-15-crown-5 (4'-nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥99.0%
4'-Nitrobenzo-15-crown-5 (4'-Nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane)

CAS Number: 60835-69-0
MDL Number: MFCD00060713
Molecular Formula: C14H19NO7
Molecular Weight: 313.31
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 98


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 2,3-(4-Nitrobenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene

Formula: C₁₄H₁₉NO₇
Melting Pt: 98 °C
MDL Number: MFCD00060713
CAS Number: 60835-69-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b27d2a537482e9834f4b8d83252dd4c8ce7a182&vid=3b27d2a537482e9834f4b8d83252dd4c8ce7a182&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR