4-Hydroxy-3-nitrobiphenyl
Supplier: TCI America
Synonyms:
2-Nitro-4-phenylphenol, 3-Nitro-[1,1'-biphenyl]-4-ol, 3-Nitrobiphenyl-4-ol
N0560-250G
N0560-25G
TCN0560-250GEA
693.62
CAD
TCN0560-250G
TCN0560-025G
4-Hydroxy-3-nitrobiphenyl
4-Hydroxy-3-nitrobiphenyl
CAS Number: 885-82-5
MDL Number: MFCD00014694
Molecular Formula: C12H9NO3
Molecular Weight: 215.21
Form: Crystal
Color: Pale Yellow
Melting point (°C): 69
Other Product Names / Synonyms: 2-Nitro-4-phenylphenol, 3-Nitrobiphenyl-4-ol
Formula:
C₁₂H₉NO₃ Melting Pt: 69 °C |
MDL Number:
MFCD00014694 CAS Number: 885-82-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...